Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl 4-bromophenyl ketone, 98%

Click to view available options
Quantity:
1g
5g
2001-29-8
C14H11BrO
275.145
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
benzyl 4-bromophenyl ketone,1-4-bromophenyl-2-phenylethanone,4'-bromo-2-phenylacetophenone,4-bromodeoxybenzoin,4-bromodesoxybenzoin,1-4-bromophenyl-2-phenyl-ethanone,ethanone, 1-4-bromophenyl-2-phenyl,1-4-bromophenyl-2-phenylethan-1-one,acmc-1chtj
519738
1-(4-bromophenyl)-2-phenylethanone
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
2001-29-8
C14H11BrO
275.145
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
benzyl 4-bromophenyl ketone,1-4-bromophenyl-2-phenylethanone,4'-bromo-2-phenylacetophenone,4-bromodeoxybenzoin,4-bromodesoxybenzoin,1-4-bromophenyl-2-phenyl-ethanone,ethanone, 1-4-bromophenyl-2-phenyl,1-4-bromophenyl-2-phenylethan-1-one,acmc-1chtj
519738
1-(4-bromophenyl)-2-phenylethanone
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
Melting Point 109°C to 112°C
Quantity 5g
Beilstein 2211322
Formula Weight 275.14
Percent Purity 98%
Chemical Name or Material Benzyl 4-bromophenyl ketone