Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 4-bromophenyl ketone, 98%, Thermo Scientific Chemicals

Catalog Number p-7026252
Click to view available options
Quantity:
1 g
5 g
2001-29-8
C14H11BrO
275.145
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
benzyl 4-bromophenyl ketone, 1-4-bromophenyl-2-phenylethanone, 4'-bromo-2-phenylacetophenone, 4-bromodeoxybenzoin, 4-bromodesoxybenzoin, 1-4-bromophenyl-2-phenyl-ethanone, ethanone, 1-4-bromophenyl-2-phenyl, 1-4-bromophenyl-2-phenylethan-1-one, acmc-1chtj
519738
1-(4-bromophenyl)-2-phenylethanone
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
2001-29-8
C14H11BrO
275.145
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
benzyl 4-bromophenyl ketone, 1-4-bromophenyl-2-phenylethanone, 4'-bromo-2-phenylacetophenone, 4-bromodeoxybenzoin, 4-bromodesoxybenzoin, 1-4-bromophenyl-2-phenyl-ethanone, ethanone, 1-4-bromophenyl-2-phenyl, 1-4-bromophenyl-2-phenylethan-1-one, acmc-1chtj
519738
1-(4-bromophenyl)-2-phenylethanone
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 109°C to 112°C
Quantity 1 g
Beilstein 2211322
Formula Weight 275.14
Percent Purity 98%
Chemical Name or Material Benzyl 4-bromophenyl ketone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit