Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl 4-bromophenyl ketone, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
2001-29-8
C14H11BrO
275.145
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
benzyl 4-bromophenyl ketone,1-4-bromophenyl-2-phenylethanone,4'-bromo-2-phenylacetophenone,4-bromodeoxybenzoin,4-bromodesoxybenzoin,1-4-bromophenyl-2-phenyl-ethanone,ethanone, 1-4-bromophenyl-2-phenyl,1-4-bromophenyl-2-phenylethan-1-one,acmc-1chtj
519738
1-(4-bromophenyl)-2-phenylethanone
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
2001-29-8
C14H11BrO
275.145
MFCD00016331
MOSIKPSTRPODHQ-UHFFFAOYSA-N
benzyl 4-bromophenyl ketone,1-4-bromophenyl-2-phenylethanone,4'-bromo-2-phenylacetophenone,4-bromodeoxybenzoin,4-bromodesoxybenzoin,1-4-bromophenyl-2-phenyl-ethanone,ethanone, 1-4-bromophenyl-2-phenyl,1-4-bromophenyl-2-phenylethan-1-one,acmc-1chtj
519738
1-(4-bromophenyl)-2-phenylethanone
C1=CC=C(C=C1)CC(=O)C2=CC=C(C=C2)Br
Quantity 1g
Color White to Yellow
Melting Point 110°C to 115°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH2COC6H4Br
Formula Weight 275.14
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Benzyl 4-bromophenyl ketone

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.