Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl acetate, 99%, Alfa Aesar™

Click to view available options
Quantity:
500g
2500g
140-11-4
C9H10O2
150.177
MFCD00008712
QUKGYYKBILRGFE-UHFFFAOYSA-N
acetic acid benzyl ester,benzyl ethanoate,acetic acid, phenylmethyl ester,phenylmethyl acetate,acetic acid, benzyl ester,alpha-acetoxytoluene,phenylmethyl ethanoate,acetoxymethyl benzene,benzylester kyseliny octove,caswell no. 081ea
8785
CHEBI:52051
benzyl acetate
CC(=O)OCC1=CC=CC=C1
140-11-4
C9H10O2
150.177
MFCD00008712
QUKGYYKBILRGFE-UHFFFAOYSA-N
acetic acid benzyl ester,benzyl ethanoate,acetic acid, phenylmethyl ester,phenylmethyl acetate,acetic acid, benzyl ester,alpha-acetoxytoluene,phenylmethyl ethanoate,acetoxymethyl benzene,benzylester kyseliny octove,caswell no. 081ea
8785
CHEBI:52051
benzyl acetate
CC(=O)OCC1=CC=CC=C1
Applications
Benzyl acetate can be used as a high boiling point solvent in coatings, like ink coating, binding agent and paint remover. It is used in soap and other chemical essences and has the effect of promote in floral and fantasy essences in jasmine, white orchid, fragrant plantain lily, gekkakou, narcissus and other essences. It is a plasticizer for ionophore membranes.

Solubility
Soluble in water 3.1 g/L.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
Odor Fruit-like
Melting Point -51°C
Flash Point 95°C (203°F)
Boiling Point 205°C to 207°C
Quantity 2500g
Assay Percent Range 99+%
Beilstein 1908121
Merck Index 14,1123
Formula Weight 150.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Benzyl acetate
Show More Show Less

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : AF5075000

TSCA : TSCA