Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Alcohol, 99%, Pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass bottle
Metal drum
100-51-6
C7H8O
108.14
MFCD00004599
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol,benzenemethanol,phenylcarbinol,benzoyl alcohol,hydroxytoluene,benzenecarbinol,phenylmethyl alcohol,alpha-toluenol,hydroxymethyl benzene,benzylalcohol
244
CHEBI:17987
phenylmethanol
C1=CC=C(C=C1)CO
100-51-6
C7H8O
108.14
MFCD00004599
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol,benzenemethanol,phenylcarbinol,benzoyl alcohol,hydroxytoluene,benzenecarbinol,phenylmethyl alcohol,alpha-toluenol,hydroxymethyl benzene,benzylalcohol
244
CHEBI:17987
phenylmethanol
C1=CC=C(C=C1)CO
Viscosity 6.6 mPa.s (20°C)
Color Colorless to Yellow
Specific Gravity 1.044
Vapor Pressure 0.13mbar at 20°C
Flash Point 96°C
Density 1.0440g/mL
Quantity 25L
Refractive Index 1.5380 to 1.5410
Packaging Metal drum
Boiling Point 205.0°C
Infrared Spectrum Authentic
Melting Point -15.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula C6H5CH2OH
Beilstein 06, 428
Merck Index 15, 1127
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: miscible with alcohol,ether,acetone,chloroform,,dmf and methylene chloride
Formula Weight 108.14
Physical Form Liquid
Percent Purity 99%
Grade Pure
Chemical Name or Material Benzyl alcohol
Show More Show Less

GHS P Statement
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.