Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Alcohol, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
4L
500mL
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

100-51-6
C7H8O
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
244
CHEBI:17987

High quality chemicals for laboratory and specialized industrial use

CAS 100-51-6
Molecular Formula C7H8O
InChI Key WVDDGKGOMKODPV-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CO
Packaging Narrow Mouth Amber Glass Bottle
IUPAC Name phenylmethanol
Molecular Weight (g/mol) 108.14
PubChem CID 244
ChEBI CHEBI:17987
Formula Weight 108.14
Specific Gravity 1L = 1.05kg
Quantity 500mL
Flash Point 93.00°C
Chemical Name or Material Benzyl Alcohol
Grade BAKER ANALYZED™ Reagent
Show More Show Less