Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Benzyl Alcohol (Certified ACS), Fisher Chemical™
Click to view available options
Quantity:
500 mL
4 L
Specifications
Boiling Range | 4°C max. |
Melting Point | -15°C |
Color | Yellow |
Density | 3.72 (Air = 1.0) |
Boiling Point | 205°C |
Flash Point | 96°C |
Ignition Residue | 0.005% max. |
Packaging | Glass Bottle |
Quantity | 500 mL |
Vapor Pressure | 0.13mbar at 20°C |
Show More |
Safety and Handling
Hazard Statement | WARNING! |
Precautionary Statement | Emergency Overview Combustible liquid. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air. NFPA Health:1 Flammability:1 Instability:0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction