Benzyl Alcohol (Certified), Fisher Chemical

Quantity 200L

 View more versions of this product

Catalog No. A396-200

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 100-51-6
    Molecular Formula C7H8O
    Formula Weight 108.14
    MDL Number MFCD00004599
    Synonym Benzenemethanol
    Chemical Name or Material Benzyl Alcohol
    Grade Certified Grade

    Specifications

    Formula Weight 108.14
    Boiling Point 205°C
    Melting Point −15°C
    Quantity 200L
    Physical Form Liquid
    Color Light Yellow
    Density 3.72 (Air = 1.0)
    Vapor Pressure 0.13mbar at 20°C
    Health Hazard 3 Emergency Overview
    Combustible liquid. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

    NFPA

    Health:1
    Flammability:1
    Instability:0
    Chlorine (Cl) Pass Test
    Additional Information Vapor Pressure: 0.13mbar at 20°C
    Identification Pass Test
    Ignition Residue 0.005% max.
    Molecular Formula C7H8O
    MDL Number MFCD00004599
    CAS 100-51-6
    Synonym Benzenemethanol
    Boiling Range 4°C max.
    Cleanup Symbol Solvent
    ChemAlert Storage Symbol Gray
    Health Hazard 2 WARNING!
    Chemical Name or Material Benzyl Alcohol
    Grade Certified Grade
       

    WARNING!

    Emergency Overview
    Combustible liquid. Irritating to eyes and skin. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

    NFPA

    Health:1
    Flammability:1
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit