Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Alcohol, Multi-Compendial, N.F., J.T.Baker™

Click to view available options
Quantity:
100mL
500mL
1L
4L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum
100-51-6
C7H8O
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol,benzenemethanol,phenylcarbinol,benzoyl alcohol,hydroxytoluene,benzenecarbinol,phenylmethyl alcohol,alpha-toluenol,hydroxymethyl benzene,benzylalcohol
244
CHEBI:17987
phenylmethanol
C1=CC=C(C=C1)CO
100-51-6
C7H8O
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
benzyl alcohol,benzenemethanol,phenylcarbinol,benzoyl alcohol,hydroxytoluene,benzenecarbinol,phenylmethyl alcohol,alpha-toluenol,hydroxymethyl benzene,benzylalcohol
244
CHEBI:17987
phenylmethanol
C1=CC=C(C=C1)CO
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL
Flash Point 93.00°C
Specific Gravity 1L = 1.05kg
Formula Weight 108.14
Grade Multi-Compendial/NF
Chemical Name or Material Benzyl Alcohol