Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Alcohol, NF, 98-100.5%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
100-51-6
C7H8O
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
OCC1=CC=CC=C1
100-51-6
C7H8O
108.14
WVDDGKGOMKODPV-UHFFFAOYSA-N
phenylmethanol
OCC1=CC=CC=C1

BE158, 100-51-6, C7H8O

Spectrum™ Benzyl Alcohol, NF is used as an anti-parasite medication used to treat head lice and as a common preservative in many injectable drugs. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Refractive Index 1.538-1.541
Quantity 4 L
CAS Max % 100%
Percent Purity 98 to 100.5%