Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Alcohol, NF, EP, bioCERTIFIED, 98-100.5%, Spectrum™ Chemical
SDP

Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
100-51-6
100-51-6

Spectrum™ Chemical bioCERTIFIED products are suitable for biopharmaceutical production.

  • Full testing including endotoxin and elemental analysis
  • Comprehensive regulatory and scientific documentation
  • Change control and batch traceability
  • Biopharm industry standard CFR 210/211 cGMP
Acidity To pass test
Identification To pass test
Packaging Amber Glass Bottle
Refractive Index 1.538-1.541
Quantity 1 L
Residue after Evaporation 0.05%
Residual Solvents To pass test
Percent Purity 98 to 100.5%
Grade NF/EP/bioCERTIFIED