Benzyl Alcohol, World/GMP, NF/BP/EP/JP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 303WORLDCS4L

Packaging Amber Glass Bottle
Quantity 4L

 View more versions of this product

Catalog No. 16-101-407

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 100-51-6
    Physical Form Liquid
    Molecular Formula C7H8O
    Formula Weight 108.14
    Synonym Benzenecarbinol
    Chemical Name or Material Benzyl Alcohol

    Specifications

    Formula Weight 108.14
    Quantity 4L
    Packaging Amber Glass Bottle
    Density 1.045g/mL at 25°C (77°F)
    Boiling Point 205°C (401°F)
    Melting Point −15°C (5°F)
    Flash Point 93°C (200°F)
    Color Clear colorless
    Physical Form Liquid
    Molecular Formula C7H8O
    CAS 100-51-6
    Synonym Benzenecarbinol
    Stability Stable under recommended storage conditions.
    Chemical Name or Material Benzyl Alcohol

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit