Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Benzoate, USP, 99-100.5%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
120-51-4
C14H12O2
212.25
SESFRYSPDFLNCH-UHFFFAOYSA-N
benzyl benzoate
O=C(OCC1=CC=CC=C1)C1=CC=CC=C1
120-51-4
C14H12O2
212.25
SESFRYSPDFLNCH-UHFFFAOYSA-N
benzyl benzoate
O=C(OCC1=CC=CC=C1)C1=CC=CC=C1

BE159, 120-51-4, C14H12O2

Spectrum™ Benzyl Benzoate, USP is used as a antiparasitic insecticide against human scabies. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 4 L
Refractive Index 1.568-1.570
CAS Max % 1
Percent Purity 99 to 100.5%