Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl bromide, 99%, Thermo Scientific Chemicals

Catalog Number AAA1353530
Click to view available options
Quantity:
50 g
250 g
1000 g
100-39-0
C7H7Br
171.037
MFCD00000172
AGEZXYOZHKGVCM-UHFFFAOYSA-N
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
7498
CHEBI:59858
bromomethylbenzene
C1=CC=C(C=C1)CBr
100-39-0
C7H7Br
171.037
MFCD00000172
AGEZXYOZHKGVCM-UHFFFAOYSA-N
benzyl bromide, bromomethyl benzene, alpha-bromotoluene, bromophenylmethane, benzene, bromomethyl, phenylmethyl bromide, 1-bromotoluene, benzylbromide, cyclite
7498
CHEBI:59858
bromomethylbenzene
C1=CC=C(C=C1)CBr

Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl bromide is widely useful in organic synthesis as a benzyl protecting group for alcohols and carboxylic acids. It is used in the preparation of foaming and frothing agents, quaternary ammonium compounds, peniclins, photographic chemicals, synthetic resins, dyes and plasticizers and esters. It is also involved in the synthesis of (E)-(2-Iodovinyl)benzene by reacting it with diiodomethane.

Solubility
Miscible with benzene, carbon tetrachloride, ethanol and ether.

Notes
Light and moisture sensitive. Incompatible with bases, alcohols, amines, oxidizing agents and corrodes steel.
TRUSTED_SUSTAINABILITY
Melting Point °C to 4°C
Density 1.438
Boiling Point 198°C to 199°C
Flash Point 87°C (189°F)
Assay Percent Range 99%
Refractive Index 1.575
Quantity 250 g
Beilstein 385801
Sensitivity Moisture sensitive
Solubility Information Miscible with benzene,carbon tetrachloride,ethanol and ether.
Formula Weight 171.04
Physical Form Liquid
Chemical Name or Material Benzyl Bromide
Show More Show Less
Hazard Category H227-H315-H319-H335
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 202-847-3
RTECSNumber XS7965000
TSCA Yes
storageNote1 Moisture sensitive
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit