Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl butyrate, 98%

Click to view available options
Quantity:
50g
250g
1000g
103-37-7
C11H14O2
178.231
MFCD00027133
VONGZNXBKCOUHB-UHFFFAOYSA-N
benzyl butyrate,benzyl n-butyrate,butanoic acid, phenylmethyl ester,phenylmethyl butanoate,benzyl n-butanoate,butyric acid, benzyl ester,phenylmethyl butyrate,benzylester kyseliny maselne,benzylbutyrate,benzyl butyrate natural
7650
benzyl butanoate
CCCC(=O)OCC1=CC=CC=C1
103-37-7
C11H14O2
178.231
MFCD00027133
VONGZNXBKCOUHB-UHFFFAOYSA-N
benzyl butyrate,benzyl n-butyrate,butanoic acid, phenylmethyl ester,phenylmethyl butanoate,benzyl n-butanoate,butyric acid, benzyl ester,phenylmethyl butyrate,benzylester kyseliny maselne,benzylbutyrate,benzyl butyrate natural
7650
benzyl butanoate
CCCC(=O)OCC1=CC=CC=C1
Quantity 1000g
Boiling Point 240°C
Flash Point 107°C (224°F)
Beilstein 2047625
Formula Weight 178.23
Percent Purity 98%
Chemical Name or Material Benzyl butyrate

RTECSNumber : ES7350000

TSCA : TSCA