Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl chloride, 99.5+%, extra pure, stabilized, ACROS Organics™

Click to view available options
Quantity:
10L
Packaging:
Plastic drum
100-44-7,75-56-9
C7H7Cl
126.59
MFCD00000889
KCXMKQUNVWSEMD-UHFFFAOYSA-N
benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol
7503
CHEBI:615597
chloromethylbenzene
C1=CC=C(C=C1)CCl
100-44-7,75-56-9
C7H7Cl
126.59
MFCD00000889
KCXMKQUNVWSEMD-UHFFFAOYSA-N
benzyl chloride,chloromethyl benzene,benzylchloride,alpha-chlorotoluene,benzylchlorid,chlorophenylmethane,benzene, chloromethyl,chlorure de benzyle,tolyl chloride,alpha-chlortoluol
7503
CHEBI:615597
chloromethylbenzene
C1=CC=C(C=C1)CCl
Alkylation reagent
Viscosity 1.280 mPa.s (20°C)
Boiling Point 179.0°C
Density 1.1000g/mL
Refractive Index 1.5370 to 1.5390
Infrared Spectrum Authentic
Specific Gravity 1.1
Flash Point 67°C
Stabilizer 0.25% propylene oxide
Packaging Plastic drum
Color Undesignated
Melting Point -39.0°C
Quantity 10L
CAS Min % 99.5
CAS Max % 100.0
Assay Percent Range 99.5% min. (GC)
Linear Formula C6H5CH2Cl
Beilstein 05,292
Merck Index 15,1132
Solubility Information Solubility in water: 0.3g/L (20°C). Other solubilities: miscible with ethanol,ethyl ether,chloroform,,acetone,slightly soluble in carbon tetrachloride
Formula Weight 126.59
Physical Form Liquid
Percent Purity 99.5+%
Grade Extra Pure
Chemical Name or Material Benzyl chloride, 99.5+%
Show More Show Less

GHS H Statement
Causes serious eye damage.
Toxic if inhaled.
May cause cancer.
May cause respiratory irritation.
Causes skin irritation.
May cause genetic defects.
May cause damage to organs through prolonged or

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautio

GHS Signal Word: Danger

EINECSNumber : 202-853-6

RTECSNumber : XS8925000

TSCA : TSCA