Alfa Aesar™ Benzyl chloride, 99+%, stab. with 0.25% propylene oxide

Manufacturer: Alfa Aesar™ 4334936

Quantity 500g

 View more versions of this product

Catalog No. AA4334936

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 100-44-7
    Molecular Formula C6H5CH2Cl
    Formula Weight 126.59
    MDL Number MFCD00000889
    UN Number UN1738
    Beilstein 471308
    Merck Index 14,1129
    Chemical Name or Material Benzyl chloride, Stabilized with 0.25% propylene oxide

    Specifications

    Formula Weight 126.59
    Health Hazard 2 GHS H Statement
    H331-H350-H373-H318-H227-H302-H315-H335
    Toxic if inhaled.
    May cause cancer.
    May cause damage to organs through prolonged or repeated exposure.
    Causes serious eye damage.
    Combustible liquid.
    Harmful if swallowed.
    Causes skin irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 100-44-7
    Beilstein 471308
    Density 1.099g/mL
    Boiling Point 178° to 180°C
    Flash Point 73°C (133°F)
    UN Number UN1738
    Merck Index 14,1129
    Health Hazard 1 Danger
    RTECS Number XS8925000
    MDL Number MFCD00000889
    Melting Point −48° to −43°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: BENZYL CHLORIDE
    Refractive Index 1.5380
    Molecular Formula C6H5CH2Cl
    Quantity 500g
    TSCA TSCA
    Physical Form Liquid
    Chemical Name or Material Benzyl chloride, Stabilized with 0.25% propylene oxide
    Assay ≥99%

    GHS H Statement
    H331-H350-H373-H318-H227-H302-H315-H335
    Toxic if inhaled.
    May cause cancer.
    May cause damage to organs through prolonged or repeated exposure.
    Causes serious eye damage.
    Combustible liquid.
    Harmful if swallowed.
    Causes skin irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit