Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl Chloride, Reagent, 99%, Spectrum™

Click to view available options
Quantity:
250 g
1 kg
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
100-44-7
C7H7Cl
126.58
KCXMKQUNVWSEMD-UHFFFAOYSA-N
(chloromethyl)benzene
ClCC1=CC=CC=C1
100-44-7
C7H7Cl
126.58
KCXMKQUNVWSEMD-UHFFFAOYSA-N
(chloromethyl)benzene
ClCC1=CC=CC=C1

B1093, 100-44-7, C6H5CH2Cl

Spectrum™ Benzyl Chloride, Reagent also known as α-chlorotoluene, is a colorless organic liquid. It is an organochlorine compound and it is widely used as a chemical building block. Benzyl chloride is used often used industrially as the precursor to benzyl esters which are used as plasticizers, flavorings, and perfumes. It is also used as a chemical intermediate in the production of dyes and many pharmaceutical products as well as a photographic developer. Benzyl chloride is also used in the synthesis of amphetamine-class drugs, because of this benzyl chloride is listed as a List II drug and its sale is monitored by the US Drug Enforcement Administration
Packaging Poly Drum
Quantity 20 L
CAS Max % 1
UN Number UN1738
Percent Purity 99%