Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl chloroacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
140-18-1
C9H9ClO2
184.62
MFCD00157062
SOGXBRHOWDEKQB-UHFFFAOYSA-N
benzyl chloroacetate,benzyl monochloracetate,acetic acid, chloro-, phenylmethyl ester,chloroacetic acid, benzyl ester,monochloroacetate,chloroacetic acid benzyl ester,benzyl-alpha-chloroacetate,phenylmethyl chloroacetate,acetic acid, chloro-, benzyl ester,phenylmethyl 2-chloroacetate
8786
benzyl 2-chloroacetate
C1=CC=C(C=C1)COC(=O)CCl
140-18-1
C9H9ClO2
184.62
MFCD00157062
SOGXBRHOWDEKQB-UHFFFAOYSA-N
benzyl chloroacetate,benzyl monochloracetate,acetic acid, chloro-, phenylmethyl ester,chloroacetic acid, benzyl ester,monochloroacetate,chloroacetic acid benzyl ester,benzyl-alpha-chloroacetate,phenylmethyl chloroacetate,acetic acid, chloro-, benzyl ester,phenylmethyl 2-chloroacetate
8786
benzyl 2-chloroacetate
C1=CC=C(C=C1)COC(=O)CCl
Flash Point >110°C
Boiling Point 132°C (12.0mmHg)
Density 1.2200g/mL
Specific Gravity 1.22
Quantity 5mL
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5CH2OC(O)CH2Cl
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 184.62
Percent Purity 98%
Chemical Name or Material Benzyl chloroacetate, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-400-0

RTECSNumber : AF8750000

TSCA : TSCA