Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl chloroformate, 97 wt%, stabilized, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
501-53-1,100-44-7
C8H7ClO2
170.6
HSDAJNMJOMSNEV-UHFFFAOYSA-N
benzyl chloroformate,carbobenzoxy chloride,benzyl chlorocarbonate,cbz chloride,benzyloxycarbonyl chloride,carbobenzyloxy chloride,carbonochloridic acid, phenylmethyl ester,benzylcarbonyl chloride,cbz-cl,chloroformic acid, benzyl ester
10387
benzyl carbonochloridate
C1=CC=C(C=C1)COC(=O)Cl
501-53-1,100-44-7
C8H7ClO2
170.6
HSDAJNMJOMSNEV-UHFFFAOYSA-N
benzyl chloroformate,carbobenzoxy chloride,benzyl chlorocarbonate,cbz chloride,benzyloxycarbonyl chloride,carbobenzyloxy chloride,carbonochloridic acid, phenylmethyl ester,benzylcarbonyl chloride,cbz-cl,chloroformic acid, benzyl ester
10387
benzyl carbonochloridate
C1=CC=C(C=C1)COC(=O)Cl

Benzyl chloroformate, 97 wt%, stabilized, Acros Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 178.0 deg.C to 180.0 deg.C, Undesignated, Melting Point: -20.0 deg.C, Molecular Weight: 170.6, Percent Purity: 97 wt%, Assay Percent Range: 96% min. (GC), Beilstein: 06,437

Benzyl chloroformate, 97 wt%, stabilized, Acros Organics, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 178.0 deg.C to 180.0 deg.C, Undesignated, Melting Point: -20.0 deg.C, Molecular Weight: 170.6, Percent Purity: 97 wt%, Assay Percent Range: 96% min. (GC), Beilstein: 06,437

Coupling reagent
Viscosity 2.36 mPa.s (20°C)
Stabilizer 0.1% anhydrous sodium carbonate
Quantity 100g
Density 1.2000g/mL
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.5160 to 1.5180
Specific Gravity 1.2
Flash Point 92°C
Melting Point -20.0°C
Boiling Point 178.0°C to 180.0°C
Color Undesignated
Assay Percent Range 96% min. (GC)
Linear Formula ClCO2CH2C6H5
Beilstein 06,437
Fieser 01,109; 02,59; 15,22
Merck Index 15,1801
Solubility Information Solubility in water: reacts
Formula Weight 170.6
Physical Form Liquid
Percent Purity 97 wt%
Chemical Name or Material Benzyl chloroformate
Show More Show Less

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 207-925-

Recommended Storage : Important Note: the material builds progressively benzyl chloride under normal storage conditions