Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl chloromethyl ether, 75%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
250g
Packaging:
Glass bottle
3587-60-8
C8H9ClO
156.61
MFCD00000886
LADPCMZCENPFGV-UHFFFAOYSA-N
benzyl chloromethyl ether,chloromethoxymethyl benzene,chloromethoxy methyl benzene,benzyloxymethyl chloride,chloromethoxymethyl-benzene,benzyl chloromethylether,benzene, chloromethoxy methyl
137983
chloromethoxymethylbenzene
C1=CC=C(C=C1)COCCl
3587-60-8
C8H9ClO
156.61
MFCD00000886
LADPCMZCENPFGV-UHFFFAOYSA-N
benzyl chloromethyl ether,chloromethoxymethyl benzene,chloromethoxy methyl benzene,benzyloxymethyl chloride,chloromethoxymethyl-benzene,benzyl chloromethylether,benzene, chloromethoxy methyl
137983
chloromethoxymethylbenzene
C1=CC=C(C=C1)COCCl
Boiling Point 102°C (14.0mmHg)
Specific Gravity 1.13
Flash Point 91°C
Packaging Glass bottle
Color Colorless to Yellow
Density 1.1300g/mL
Quantity 50g
Infrared Spectrum Authentic
CAS Min % 70.0
CAS Max % 100.0
Assay Percent Range 70% min. (GC)
Linear Formula C6H5CH2OCH2Cl
Formula Weight 156.61
Physical Form Liquid
Percent Purity 75%
Chemical Name or Material Benzyl chloromethyl ether
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause cancer.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger