Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl cinnamate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
103-41-3
C16H14O2
238.29
MFCD00004789
NGHOLYJTSCBCGC-QXMHVHEDSA-N
benzyl alcohol cinnamic ester,benzylcinnamate,benzyl alcohol, cinnamate,wln: r1u1vo1r,benzyl .gamma.-phenylacrylate,benzyl cis-cinnamate,benzyl z-cinnamate,2-propenoic acid, 3-phenyl-, phenylmethyl ester,z-benzyl 3-phenylacrylate,cinnamic acid, benzyl ester, z
15558051
benzyl (Z)-3-phenylprop-2-enoate
C1=CC=C(C=C1)COC(=O)C=CC2=CC=CC=C2
103-41-3
C16H14O2
238.29
MFCD00004789
NGHOLYJTSCBCGC-QXMHVHEDSA-N
benzyl alcohol cinnamic ester,benzylcinnamate,benzyl alcohol, cinnamate,wln: r1u1vo1r,benzyl .gamma.-phenylacrylate,benzyl cis-cinnamate,benzyl z-cinnamate,2-propenoic acid, 3-phenyl-, phenylmethyl ester,z-benzyl 3-phenylacrylate,cinnamic acid, benzyl ester, z
15558051
benzyl (Z)-3-phenylprop-2-enoate
C1=CC=C(C=C1)COC(=O)C=CC2=CC=CC=C2
Melting Point 33°C to 37°C
Packaging Glass bottle
Refractive Index 1.4025 to 1.4045
Flash Point >112°C
Boiling Point 195°C to 200°C (5.0mmHg)
Quantity 500g
Infrared Spectrum Authentic
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5CH=CHCO2CH2C6H5
Beilstein 09, 584
Merck Index 15, 1133
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol,ether and oils,practically insoluble in propylene glycol and,glycerin
Formula Weight 238.29
Physical Form Crystalline Mass or Liquid After Melting
Percent Purity 99%
Chemical Name or Material Benzyl cinnamate, 99%
Show More Show Less

EINECSNumber : 203-109-3

RTECSNumber : GD8400000

TSCA : TSCA