Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl ethyl malonate, 96%

Manufacturer:  Alfa Aesar™ H3112606

Quantity 5g

 View more versions of this product

Catalog No. AAH3112606

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

42998-51-6
222.24
Malonic acid benzyl ethyl ester
C12H14O4
MFCD00009194
Benzyl ethyl malonate

Specifications

42998-51-6
C12H14O4
MFCD00009194
Benzyl ethyl malonate
5g
222.24
Malonic acid benzyl ethyl ester
96%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit