Alfa Aesar™ Benzyl ethyl malonate, 96%

Manufacturer: Alfa Aesar™ H3112614

Quantity 25g

 View more versions of this product

Catalog No. AAH3112614

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 42998-51-6
    Molecular Formula C12H14O4
    Formula Weight 222.24
    MDL Number MFCD00009194
    Synonym Malonic acid benzyl ethyl ester
    Chemical Name or Material Benzyl ethyl malonate

    Specifications

    Formula Weight 222.24
    Quantity 25g
    Molecular Formula C12H14O4
    MDL Number MFCD00009194
    CAS 42998-51-6
    Synonym Malonic acid benzyl ethyl ester
    Chemical Name or Material Benzyl ethyl malonate
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit