Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl ethyl malonate, tech. 85%

Manufacturer:  Alfa Aesar™ B2301106

Quantity 5g

 View more versions of this product

Catalog No. AAB2301106


  Check Availability
Add to cart

Specifications

Description

Applications
Benzyl ethyl malonate is used as a pharmaceutical intermediate.

Solubility
Soluble in water 525.7 mg/L at 25°C. Soluble in alcohol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

42998-51-6
222.24
Malonic acid benzyl ethyl ester
Technical Grade
C12H14O4
MFCD00009194
Benzyl ethyl malonate

Specifications

42998-51-6
222.24
Malonic acid benzyl ethyl ester
5g
85%
C12H14O4
MFCD00009194
145°C (4mmHg)
Benzyl ethyl malonate
Technical Grade
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit