Alfa Aesar™ Benzyl ethyl malonate, tech. 85%

Manufacturer: Alfa Aesar™ B2301122

Quantity 100g

 View more versions of this product

Catalog No. AAB2301122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 42998-51-6
    Molecular Formula C12H14O4
    Formula Weight 222.24
    MDL Number MFCD00009194
    Synonym Malonic acid benzyl ethyl ester
    Chemical Name or Material Benzyl ethyl malonate
    Grade Technical Grade

    Specifications

    Formula Weight 222.24
    Molecular Formula C12H14O4
    MDL Number MFCD00009194
    CAS 42998-51-6
    Synonym Malonic acid benzyl ethyl ester
    Boiling Point 145°C (4mmHg)
    Quantity 100g
    Chemical Name or Material Benzyl ethyl malonate
    Assay 85%
    Grade Technical Grade

    Description

    Applications
    Benzyl ethyl malonate is used as a pharmaceutical intermediate.

    Solubility
    Soluble in water 525.7 mg/L at 25°C. Soluble in alcohol.

    Notes
    Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit