Alfa Aesar™ Benzyl glycinate hydrochloride, 98%

Manufacturer: Alfa Aesar™ H5024706

Quantity 5g

 View more versions of this product

Catalog No. AAH5024706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2462-31-9
    Molecular Formula C9H11NO2·HCl
    Linear Formula H2NCH2CO2CH2C6H5·HCl
    Formula Weight 201.65
    MDL Number MFCD00035442
    Synonym H-Gly-OBzl·HCl
    Chemical Name or Material Benzyl glycinate hydrochloride

    Specifications

    Formula Weight 201.65
    Molecular Formula C9H11NO2·HCl
    Linear Formula H2NCH2CO2CH2C6H5·HCl
    MDL Number MFCD00035442
    CAS 2462-31-9
    Synonym H-Gly-OBzl·HCl
    Melting Point 139° to 140°C
    Quantity 5g
    Recommended Storage 2° to 8°C
    Chemical Name or Material Benzyl glycinate hydrochloride
    Assay 98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit