Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl isovalerate, 98%

Click to view available options
Quantity:
50g
250g
103-38-8
C12H16O2
192.258
MFCD00048348
HVJKZICIMIWFCP-UHFFFAOYSA-N
benzyl isovalerate,benzyl 3-methylbutyrate,benzyl isopentanoate,butanoic acid, 3-methyl-, phenylmethyl ester,isovaleric acid, benzyl ester,phenylmethyl 3-methylbutanoate,benzyl 3-methyl butyrate,benzyl isovalerianate,isopropyl acetic acid, benzyl ester,isopentanoic acid, phenylmethyl ester
7651
benzyl 3-methylbutanoate
CC(C)CC(=O)OCC1=CC=CC=C1
103-38-8
C12H16O2
192.258
MFCD00048348
HVJKZICIMIWFCP-UHFFFAOYSA-N
benzyl isovalerate,benzyl 3-methylbutyrate,benzyl isopentanoate,butanoic acid, 3-methyl-, phenylmethyl ester,isovaleric acid, benzyl ester,phenylmethyl 3-methylbutanoate,benzyl 3-methyl butyrate,benzyl isovalerianate,isopropyl acetic acid, benzyl ester,isopentanoic acid, phenylmethyl ester
7651
benzyl 3-methylbutanoate
CC(C)CC(=O)OCC1=CC=CC=C1
Quantity 250g
Boiling Point 245°C
Formula Weight 192.26
Percent Purity 98%
Chemical Name or Material Benzyl isovalerate

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

TSCA : TSCA