Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl methyl ether, 99%

Click to view available options
Quantity:
10g
50g
250g
538-86-3
C8H10O
122.167
MFCD00025901
GQKZBCPTCWJTAS-UHFFFAOYSA-N
benzyl methyl ether,methoxymethyl benzene,methyl benzyl ether,benzene, methoxymethyl,ether, benzyl methyl,benzylmethylether,unii-f22rls78bd,.alpha.-methoxytoluene,f22rls78bd,methoxyphenylmethane
10869
methoxymethylbenzene
COCC1=CC=CC=C1
538-86-3
C8H10O
122.167
MFCD00025901
GQKZBCPTCWJTAS-UHFFFAOYSA-N
benzyl methyl ether,methoxymethyl benzene,methyl benzyl ether,benzene, methoxymethyl,ether, benzyl methyl,benzylmethylether,unii-f22rls78bd,.alpha.-methoxytoluene,f22rls78bd,methoxyphenylmethane
10869
methoxymethylbenzene
COCC1=CC=CC=C1
Applications
Benzyl methyl ether is used to produce benzaldehyde. It is used as perfuming agents.

Solubility
Insoluble in water.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Quantity 250g
Flash Point 56°C (133°F)
Melting Point -53°C
Boiling Point 173°C to 175°C
UN Number UN3271
Merck Index 14,1140
Formula Weight 122.17
Percent Purity 99%
Chemical Name or Material Benzyl methyl ether

GHS H Statement
H226
Flammable liquid and vapour.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S.

TSCA : TSCA