Learn More
Benzyl n-butyl phthalate, 98%, Thermo Scientific Chemicals
Description
Benzyl n-butyl phthalate is used as a plasticizer for vinyl foams. It is also used in floor tiles, in traffic cones, food conveyor belts and artificial leather. Further, it acts as an organic intermediate. In addition to this, it is used as a perfume fixative.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzyl n-butyl phthalate is used as a plasticizer for vinyl foams. It is also used in floor tiles, in traffic cones, food conveyor belts and artificial leather. Further, it acts as an organic intermediate. In addition to this, it is used as a perfume fixative.
Solubility
Slightly miscible with water.
Notes
Store in a cool place. Incompatible with strong oxidizing agents, alkalis and strong bases.
Specifications
Density | 1.1 |
Boiling Point | 370°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.54 |
Quantity | 100 g |
UN Number | UN3082 |
Solubility Information | Slightly miscible with water. |
Formula Weight | 312.37 |
Percent Purity | 98% |
Chemical Name or Material | Benzyl n-butyl phthalate |
Safety and Handling
Hazard Category | H319-H360 |
Hazard Statement | GHS H Statement H360 May damage fertility or the unborn child. |
Precautionary Statement | P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c |
DOTInformation | Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S. |
EINECSNumber | 201-622-7 |
TSCA | Yes |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.