Learn More
Benzyl phenyl ether, 97%, Thermo Scientific Chemicals
Description
Benzyl phenyl ether reacts with aluminum bromide in chlorobenzene solution to afford a mixture of phenol, o-benzyl phenol and dichlorodiphenylmethane. It is a useful as model compound in catalytic chemistry to represent the a-O4 ether bond in lignin and coal. It contains a weak ether bond of 234kJ/mol and belongs to the most thermo-labile compounds in lignin and low rank coal. Influence of alkali carbonates, common additives in biomass conversion, on the reaction pathways of BPE in superheated water has been reported. Cesium-exchanged heteropolyacid catalyzed decomposition of benzyl phenyl ether to aromatics has been investigated. Photo-Claisen rearrangements of benzyl phenyl ether was investigated in cation-exchanged Y zeolites and polyethylenes of differing crystallinities.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzyl phenyl ether reacts with aluminum bromide in chlorobenzene solution to afford a mixture of phenol, o-benzyl phenol and dichlorodiphenylmethane. It is a useful as model compound in catalytic chemistry to represent the a-O4 ether bond in lignin and coal. It contains a weak ether bond of 234kJ/mol and belongs to the most thermo-labile compounds in lignin and low rank coal. Influence of alkali carbonates, common additives in biomass conversion, on the reaction pathways of BPE in superheated water has been reported. Cesium-exchanged heteropolyacid catalyzed decomposition of benzyl phenyl ether to aromatics has been investigated. Photo-Claisen rearrangements of benzyl phenyl ether was investigated in cation-exchanged Y zeolites and polyethylenes of differing crystallinities.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Melting Point | 37°C to 40°C |
Boiling Point | 286°C to 287°C |
Flash Point | >110°C (230°F) |
Quantity | 25 g |
Beilstein | 2045713 |
Solubility Information | Insoluble in water. |
Formula Weight | 184.24 |
Percent Purity | 97% |
Chemical Name or Material | Benzyl phenyl ether |
Safety and Handling
Hazard Category | H315-H319-H335 |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.