Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl phenyl sulfone, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
3112-88-7
C13H12O2S
232.297
MFCD00025040
FABCMLOTUSCWOR-UHFFFAOYSA-N
benzyl phenyl sulfone,benzylsulfonyl benzene,sulfone, benzyl phenyl,benzylphenylsulfone,phenyl benzyl sulfone,benzyl phenyl sulphone,benzene, phenylmethyl sulfonyl,phenylmethyl sulfonyl benzene,phenylmethyl sulphonyl benzene,benzenesulfonyl methyl benzene
76561
benzenesulfonylmethylbenzene
C1=CC=C(C=C1)CS(=O)(=O)C2=CC=CC=C2
3112-88-7
C13H12O2S
232.297
MFCD00025040
FABCMLOTUSCWOR-UHFFFAOYSA-N
benzyl phenyl sulfone,benzylsulfonyl benzene,sulfone, benzyl phenyl,benzylphenylsulfone,phenyl benzyl sulfone,benzyl phenyl sulphone,benzene, phenylmethyl sulfonyl,phenylmethyl sulfonyl benzene,phenylmethyl sulphonyl benzene,benzenesulfonyl methyl benzene
76561
benzenesulfonylmethylbenzene
C1=CC=C(C=C1)CS(=O)(=O)C2=CC=CC=C2
Quantity 10g
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
Melting Point 146°C to 150°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5CH2SO2C6H5
Formula Weight 232.3
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Benzyl phenyl sulfone
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.