Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl propargyl ether, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
4039-82-1
C10H10O
146.19
MFCD00483987
PAQVEXAFKDWGOT-UHFFFAOYSA-N
benzyl propargyl ether,benzylpropargyl ether,1-benzyloxy-2-propyne,prop-2-yn-1-yloxy methyl benzene,benzene, 2-propynyloxy methyl,prop-2-ynyloxy methyl benzene,propargylbenzyl ether,benzyloxy prop-2-yne
6917484
prop-2-ynoxymethylbenzene
C#CCOCC1=CC=CC=C1
4039-82-1
C10H10O
146.19
MFCD00483987
PAQVEXAFKDWGOT-UHFFFAOYSA-N
benzyl propargyl ether,benzylpropargyl ether,1-benzyloxy-2-propyne,prop-2-yn-1-yloxy methyl benzene,benzene, 2-propynyloxy methyl,prop-2-ynyloxy methyl benzene,propargylbenzyl ether,benzyloxy prop-2-yne
6917484
prop-2-ynoxymethylbenzene
C#CCOCC1=CC=CC=C1
Boiling Point 100°C to 101°C (18.0mmHg)
Specific Gravity 1.011
Refractive Index 1.5190 to 1.521
Density 1.0110g/mL
Quantity 1g
Color Colorless to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Linear Formula C6H5CH2OCH2C∽CH
Formula Weight 146.19
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Benzyl propargyl ether
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning