Alfa Aesar™ Benzyl propionate, 99%

Manufacturer: Alfa Aesar™ B231860B

Quantity 1000g

 View more versions of this product

Catalog No. AAB231860B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 122-63-4
    Molecular Formula CH3CH2CO2CH2C6H5
    Formula Weight 164.21
    MDL Number MFCD00027003
    Beilstein 2046122
    Synonym Propionic acid benzyl ester
    Chemical Name or Material Benzyl propionate

    Specifications

    Formula Weight 164.21
    TSCA TSCA
    Molecular Formula CH3CH2CO2CH2C6H5
    Melting Point 221° to 223°C
    MDL Number MFCD00027003
    Beilstein 2046122
    Boiling Point 222°C
    Quantity 1000g
    Flash Point 101°C (213°F)
    Synonym Propionic acid benzyl ester
    RTECS Number UA2537603
    CAS 122-63-4
    Chemical Name or Material Benzyl propionate
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit