Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl propionate, 99%

Click to view available options
Quantity:
50g
250g
122-63-4
C10H12O2
164.204
MFCD00027003
VHOMAPWVLKRQAZ-UHFFFAOYSA-N
benzyl propionate,propanoic acid, phenylmethyl ester,propionic acid, benzyl ester,phenylmethyl propanoate,propionic acid benzyl ester,phenylmethyl propionate,benzyl propionate natrual,benzyl n-propionate,fema no. 2150,propionic acid, benzyl ester 6ci,7ci,8ci
31219
benzyl propanoate
CCC(=O)OCC1=CC=CC=C1
122-63-4
C10H12O2
164.204
MFCD00027003
VHOMAPWVLKRQAZ-UHFFFAOYSA-N
benzyl propionate,propanoic acid, phenylmethyl ester,propionic acid, benzyl ester,phenylmethyl propanoate,propionic acid benzyl ester,phenylmethyl propionate,benzyl propionate natrual,benzyl n-propionate,fema no. 2150,propionic acid, benzyl ester 6ci,7ci,8ci
31219
benzyl propanoate
CCC(=O)OCC1=CC=CC=C1
Melting Point 221°C to 223°C
Boiling Point 222°C
Quantity 50g
Flash Point 101°C (213°F)
Beilstein 2046122
Formula Weight 164.21
Percent Purity 99%
Chemical Name or Material Benzyl propionate

RTECSNumber : UA2537603

TSCA : TSCA