Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl sulfide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
538-74-9
C14H14S
214.3
MFCD00004784
LUFPJJNWMYZRQE-UHFFFAOYSA-N
dibenzyl sulfide,benzyl sulfide,dibenzyl sulphide,benzyl thioether,benzyl monosulfide,dibenzyl monosulfide,dibenzyl thioether,sulfide, dibenzyl,1,3-diphenyl-2-thiapropane,benzene, 1,1'-thiobis methylene bis
10867
benzylsulfanylmethylbenzene
C1=CC=C(C=C1)CSCC2=CC=CC=C2
538-74-9
C14H14S
214.3
MFCD00004784
LUFPJJNWMYZRQE-UHFFFAOYSA-N
dibenzyl sulfide,benzyl sulfide,dibenzyl sulphide,benzyl thioether,benzyl monosulfide,dibenzyl monosulfide,dibenzyl thioether,sulfide, dibenzyl,1,3-diphenyl-2-thiapropane,benzene, 1,1'-thiobis methylene bis
10867
benzylsulfanylmethylbenzene
C1=CC=C(C=C1)CSCC2=CC=CC=C2
Quantity 100g
Color White
Flash Point >110°C
Melting Point 44°C to 49°C
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Merck Index 15, 1147
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether
Formula Weight 214.3
Physical Form Crystals
Percent Purity ≥97.5%
Chemical Name or Material Benzyl sulfide, 98%
Show More Show Less

STENCH

EINECSNumber : 208-703-6

RTECSNumber : DC0512500

TSCA : TSCA