Alfa Aesar™ Benzyl thiocyanate, 98+%

Manufacturer: Alfa Aesar™ B2390218

Quantity 50g

 View more versions of this product

Catalog No. AAB2390218

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3012-37-1
    Molecular Formula C6H5CH2SCN
    Formula Weight 149.22
    MDL Number MFCD00001832
    UN Number UN2811
    Beilstein 1859726
    Synonym Benzyl rhodanide
    Chemical Name or Material Benzyl thiocyanate

    Specifications

    Formula Weight 149.22
    Melting Point 40° to 43°C
    UN Number UN2811
    Quantity 50g
    Odor Irritating
    Health Hazard 2 GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
    RTECS Number XK8155000
    TSCA TSCA
    Molecular Formula C6H5CH2SCN
    MDL Number MFCD00001832
    CAS 3012-37-1
    Beilstein 1859726
    Boiling Point 230°C
    Flash Point 110°C (230°F)
    Synonym Benzyl rhodanide
    Health Hazard 1 Danger
    Chemical Name or Material Benzyl thiocyanate
    Assay ≥98%

    GHS H Statement
    H331-H302-H312
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P261-P280-P304+P340-P311-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit