Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyl thiocyanate, 98+%

Click to view available options
Quantity:
250g
3012-37-1
C8H7NS
149.211
MFCD00001832
ABNDFSOIUFLJAH-UHFFFAOYSA-N
tropeolin,thiocyanic acid, phenylmethyl ester,solvat 14,benzylthiocyanate,benzylrhodonid,thiocyanic acid, benzyl ester,alpha-thiocyanatotoluene,benzyl-thiocyanate,thiocyanic acid benzyl ester,phenylmethyl thiocyanate
18170
CHEBI:16017
benzyl thiocyanate
C1=CC=C(C=C1)CSC#N
3012-37-1
C8H7NS
149.211
MFCD00001832
ABNDFSOIUFLJAH-UHFFFAOYSA-N
tropeolin,thiocyanic acid, phenylmethyl ester,solvat 14,benzylthiocyanate,benzylrhodonid,thiocyanic acid, benzyl ester,alpha-thiocyanatotoluene,benzyl-thiocyanate,thiocyanic acid benzyl ester,phenylmethyl thiocyanate
18170
CHEBI:16017
benzyl thiocyanate
C1=CC=C(C=C1)CSC#N
Odor Irritating
Quantity 250g
Flash Point 110°C (230°F)
Boiling Point 230°C
Melting Point 40°C to 43°C
UN Number UN2811
Beilstein 1859726
Formula Weight 149.22
Percent Purity ≥98%
Chemical Name or Material Benzyl thiocyanate

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : XK8155000

TSCA : TSCA