Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyl (triphenylphosphoranylidene)acetate, 97%, Thermo Scientific Chemicals

Catalog Number AC377220050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
15097-38-8
C27H23O2P
410.45
MFCD00191787
INKMLGJBBDRIQR-UHFFFAOYSA-N
benzyl 2-triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene-acetate, benzyl 2-triphenyl-??-phosphanylidene acetate, acetic acid, triphenylphosphoranylidene-, phenylmethyl ester, phenylmethyl 2-triphenylylidene acetate, benzyl 2-triphenyl, acmc-20ah9g, ksc526q3l
3862746
benzyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
O=C(OCC1=CC=CC=C1)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
15097-38-8
C27H23O2P
410.45
MFCD00191787
INKMLGJBBDRIQR-UHFFFAOYSA-N
benzyl 2-triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene acetate, benzyl triphenylphosphoranylidene-acetate, benzyl 2-triphenyl-??-phosphanylidene acetate, acetic acid, triphenylphosphoranylidene-, phenylmethyl ester, phenylmethyl 2-triphenylylidene acetate, benzyl 2-triphenyl, acmc-20ah9g, ksc526q3l
3862746
benzyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
O=C(OCC1=CC=CC=C1)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 120.0°C to 127.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCOOCH2C6H5
Quantity 5 g
Formula Weight 410.45
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Benzyl (triphenylphosphoranylidene)acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit