Alfa Aesar™ Benzyldimethylhexadecylammonium chloride hydrate, 95%

Manufacturer: Alfa Aesar™ 3237709

Quantity 10g

 View more versions of this product

Catalog No. AA3237709

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 122-18-9
    Linear Formula CH3(CH2)15N(CH2C6H5)(CH3)2Cl·xH2O
    Formula Weight 396.10 (Anhydrous)
    MDL Number MFCD00011625
    UN Number UN3261
    Beilstein 3802341
    Merck Index 14,2017
    Synonym Benzylcetyldimethylammonium chloride hydrate; Cetalkonium chloride
    Chemical Name or Material Benzyldimethylhexadecylammonium chloride hydrate

    Specifications

    Formula Weight 396.10 (Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3261
    Beilstein 3802341
    Merck Index 14,2017
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Physical Form Powder
    MDL Number MFCD00011625
    Synonym Benzylcetyldimethylammonium chloride hydrate; Cetalkonium chloride
    Melting Point 54° to 58°C
    Health Hazard 1 Danger
    Quantity 10g
    TSCA TSCA
    Linear Formula CH3(CH2)15N(CH2C6H5)(CH3)2Cl·xH2O
    CAS 122-18-9
    Color White
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    RTECS Number BO6822450
    Chemical Name or Material Benzyldimethylhexadecylammonium chloride hydrate
    Assay 95%

    GHS H Statement
    H301-H311-H314-H318
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit