Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyldiphenylphosphine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
7650-91-1
C19H17P
276.319
MFCD00014083
UZCPNEBHTFYJNY-UHFFFAOYSA-N
benzyldiphenylphosphine,phosphine, diphenyl phenylmethyl,phosphine, benzyldiphenyl,diphenylbenzylphosphine,benzyl diphenyl phosphane,benzyl-diphenylphosphine,benzyl-diphenyl-phosphane,di phenyl-phenylmethyl phosphane,diphenylphosphionomethyl polystyrene,benzyldiphenylphosphine, polymer-bound
603920
benzyl(diphenyl)phosphane
C1=CC=C(C=C1)CP(C2=CC=CC=C2)C3=CC=CC=C3
7650-91-1
C19H17P
276.319
MFCD00014083
UZCPNEBHTFYJNY-UHFFFAOYSA-N
benzyldiphenylphosphine,phosphine, diphenyl phenylmethyl,phosphine, benzyldiphenyl,diphenylbenzylphosphine,benzyl diphenyl phosphane,benzyl-diphenylphosphine,benzyl-diphenyl-phosphane,di phenyl-phenylmethyl phosphane,diphenylphosphionomethyl polystyrene,benzyldiphenylphosphine, polymer-bound
603920
benzyl(diphenyl)phosphane
C1=CC=C(C=C1)CP(C2=CC=CC=C2)C3=CC=CC=C3
Infrared Spectrum Authentic
Quantity 1g
Melting Point 74.0°C to 75.0°C
Color White
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H5CH2P(C6H5)2
Solubility Information Solubility in water: insoluble.
Formula Weight 276.32
Physical Form Powder
Percent Purity 97%
Chemical Name or Material Benzyldiphenylphosphine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.