Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylmagnesium chloride, 1.5M solution in THF, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
6921-34-2,109-99-9
C7H7ClMg
150.888
MFCD00000469
SCEZYJKGDJPHQO-UHFFFAOYSA-M
benzylmagnesium chloride,magnesium, chloro phenylmethyl,phch2mgcl,benzyl chloro magnesium,benzyl magnesium chloride,chloro phenylmethyl magnesium,benzylmagnesiumchloride,bnmgcl,benzyl-magnesium chloride,chloro phenylmethyl-magnesium
2733352
magnesium;methanidylbenzene;chloride
[CH2-]C1=CC=CC=C1.[Mg+2].[Cl-]
6921-34-2,109-99-9
C7H7ClMg
150.888
MFCD00000469
SCEZYJKGDJPHQO-UHFFFAOYSA-M
benzylmagnesium chloride,magnesium, chloro phenylmethyl,phch2mgcl,benzyl chloro magnesium,benzyl magnesium chloride,chloro phenylmethyl magnesium,benzylmagnesiumchloride,bnmgcl,benzyl-magnesium chloride,chloro phenylmethyl-magnesium
2733352
magnesium;methanidylbenzene;chloride
[CH2-]C1=CC=CC=C1.[Mg+2].[Cl-]
CAS Min % 20.51,75.83
CAS Max % 24.17,79.49
Appearance of Solution Clear solution
Linear Formula (C6H5)CH2MgCl
Concentration 1.40 to 1.65M
Density 1.0300g/mL
Formula Weight 150.89
Physical Form Iridescent Powder or Crystals
Percent Purity 19 to 21% (as BMC) (Total base)
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: reacts.
Specific Gravity 1.03
Color Brown to Black
Quantity 800mL
Flash Point −17°C
Chemical Name or Material Benzylmagnesium chloride, 1.5M solution in THF
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Suspected of causing cancer. May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

Danger