Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals

Catalog Number p-7033951
Click to view available options
Quantity:
100 mL
6921-34-2
C7H7ClMg
150.89
MFCD00000469
NRAFPLGJPPJUNB-UHFFFAOYSA-M
benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium
2733352
magnesium;methanidylbenzene;chloride
Cl[Mg]CC1=CC=CC=C1
6921-34-2
C7H7ClMg
150.89
MFCD00000469
NRAFPLGJPPJUNB-UHFFFAOYSA-M
benzylmagnesium chloride, magnesium, chloro phenylmethyl, phch2mgcl, benzyl chloro magnesium, benzyl magnesium chloride, chloro phenylmethyl magnesium, benzylmagnesiumchloride, bnmgcl, benzyl-magnesium chloride, chloro phenylmethyl-magnesium
2733352
magnesium;methanidylbenzene;chloride
Cl[Mg]CC1=CC=CC=C1

Benzyl magnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is also used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl magnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is also used as organic chemical synthesis intermediate.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Solubility Information It reacts violently with water.
Formula Weight 150.89
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material Benzylmagnesium chloride
Hazard Category H225-H302-H314-H335
Hazard Statement GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit