Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester, 97%

Click to view available options
Quantity:
1g
5g
25g
88568-95-0
C13H18NO7P
331.261
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate,+/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester,n-cbz-2-phosphonoglycine trimethyl ester,n-benzyloxy carbonyl,+/--cbz-alpha-phosphonoglycine trimethyl ester,methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate,+/--n-cbz-alpha-phosphonoglycine trimethyl ester,z-alpha-phosphonoglycine trimethyl ester,cbz-alpha-phosphono-dl-glycine trimethyl ester
2734718
methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
88568-95-0
C13H18NO7P
331.261
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate,+/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester,n-cbz-2-phosphonoglycine trimethyl ester,n-benzyloxy carbonyl,+/--cbz-alpha-phosphonoglycine trimethyl ester,methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate,+/--n-cbz-alpha-phosphonoglycine trimethyl ester,z-alpha-phosphonoglycine trimethyl ester,cbz-alpha-phosphono-dl-glycine trimethyl ester
2734718
methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
Applications
(±)-Z-α-Phosphonoglycine trimethyl ester is the starting material for the synthesis of (Z)-dehydroamino acids by Wittig-Horner reaction with aldehydes, the dehydroamino acids can be stereo selectively reduced to amino acids.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Melting Point 76°C to 80°C
Quantity 1g
Formula Weight 331.27
Percent Purity 97%
Chemical Name or Material (+/-)-Benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning