Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Benzyltri-n-butylammonium bromide, 99%
Click to view available options
Quantity:
25g
50g
250g
Description
Applications
Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature.
Benzyltri-n-butylammonium bromide will react with N-vinyl-phthalimide to produce N-trans-cinnamyl-phthalimide.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature.
Specifications
Quantity | 250g |
Melting Point | ~175°C (decomposition) |
Sensitivity | Hygroscopic |
Beilstein | 3776294 |
Solubility Information | Soluble in water. |
Formula Weight | 356.4 |
Percent Purity | 99% |
Chemical Name or Material | Benzyltri-n-butylammonium bromide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 246-819-9
TSCA : No
Recommended Storage : Ambient temperatures