Alfa Aesar™ Benzyltri-n-butylammonium chloride, 98%

Manufacturer: Alfa Aesar™ A1034522

Quantity 100g

 View more versions of this product

Catalog No. AAA1034522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 23616-79-7
    Molecular Formula [CH3(CH2)3]3N(CH2C6H5)Cl
    Formula Weight 311.94
    MDL Number MFCD00011849
    Beilstein 3776210
    Chemical Name or Material Benzyltri-n-butylammonium chloride

    Specifications

    Formula Weight 311.94
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula [CH3(CH2)3]3N(CH2C6H5)Cl
    CAS 23616-79-7
    Beilstein 3776210
    Health Hazard 2 GHS H Statement
    H301-H318
    Toxic if swallowed.
    Causes serious eye damage.
    MDL Number MFCD00011849
    Health Hazard 1 Danger
    Quantity 100g
    TSCA TSCA
    Melting Point 156° to 162°C
    Chemical Name or Material Benzyltri-n-butylammonium chloride
    Assay 98%

    GHS H Statement
    H301-H318
    Toxic if swallowed.
    Causes serious eye damage.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P310-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit