Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltributylammonium chloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Glass bottle
23616-79-7
C19H34ClN
311.94
MFCD00011849
VJGNLOIQCWLBJR-UHFFFAOYSA-M
benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
159952
benzyl(tributyl)azanium;chloride
[Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1
23616-79-7
C19H34ClN
311.94
MFCD00011849
VJGNLOIQCWLBJR-UHFFFAOYSA-M
benzyltributylammonium chloride, n-benzyl-n,n-dibutylbutan-1-aminium chloride, benzyltri-n-butylammonium chloride, benzyl tributyl ammonium chloride, benzenemethanaminium, n,n,n-tributyl-, chloride, tributylbenzylammonium chloride, benzyltributylazanium chloride, benzyltributyl ammonium chloride, benzyl-tributyl-azanium, btbac
159952
benzyl(tributyl)azanium;chloride
[Cl-].CCCC[N+](CCCC)(CCCC)CC1=CC=CC=C1
Phase transfer catalyst
Melting Point 155°C to 163°C
Color White to Ivory
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98+%
Linear Formula C6H5CH2N[(CH2)3CH3]3Cl
Fieser 06,41
Solubility Information Solubility in water: freely soluble in water
Formula Weight 311.94
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material Benzyltributylammonium chloride, 98+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 245-787-3

TSCA : TSCA