Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Benzyltriethylammonium borohydride, 96%

Click to view available options
Quantity:
5g
25g
100g
85874-45-9
C13H22BN
203.136
MFCD00191785
HQGWCEJYXHFBSA-UHFFFAOYSA-N
benzyltriethylammonium borohydride,benzyltriethylazanium ? 1-boranuidylidene,n,n,n-triethyl-benzenemethanaminium tetrahydroborate
10899792
benzyl(triethyl)azanium;boron(1-)
[B-].CC[N+](CC)(CC)CC1=CC=CC=C1
85874-45-9
C13H22BN
203.136
MFCD00191785
HQGWCEJYXHFBSA-UHFFFAOYSA-N
benzyltriethylammonium borohydride,benzyltriethylazanium ? 1-boranuidylidene,n,n,n-triethyl-benzenemethanaminium tetrahydroborate
10899792
benzyl(triethyl)azanium;boron(1-)
[B-].CC[N+](CC)(CC)CC1=CC=CC=C1
Applications
Benzyltriethylammonium borohydride involves in the conversion of alkenes to the anti-Markovnikov alcohols in presence of TMS chloride or TiCl4. It combines with TMS chloride, selectively reduces carboxylic acids to alcohols; esters, lactones, nitro- and chloro-substituents are unaffected. It is used as reducing reagent.

Solubility
Souble in Chloroform.

Notes
Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.
Quantity 25g
Melting Point ~146°C (decomposition)
Linear Formula (CH3CH2)3N(CH2C6H5)BH4
UN Number UN3131
Formula Weight 207.17
Percent Purity 96%
Chemical Name or Material Benzyltriethylammonium borohydride

GHS H Statement
H261-H314-H318-H302
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.