Alfa Aesar™ Benzyltriethylammonium borohydride, 96%

Manufacturer: Alfa Aesar™ B2495222

Quantity 100g

 View more versions of this product

Catalog No. AAB2495222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85874-45-9
    Molecular Formula C13H26BN
    Linear Formula (CH3CH2)3N(CH2C6H5)BH4
    Formula Weight 207.17
    MDL Number MFCD00191785
    UN Number UN3131
    Synonym Benzyltriethylammonium tetrahydridoborate
    Chemical Name or Material Benzyltriethylammonium borohydride

    Specifications

    Formula Weight 207.17
    Molecular Formula C13H26BN
    Linear Formula (CH3CH2)3N(CH2C6H5)BH4
    MDL Number MFCD00191785
    UN Number UN3131
    Melting Point ≈146°C (dec.)
    Quantity 100g
    CAS 85874-45-9
    Synonym Benzyltriethylammonium tetrahydridoborate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H261-H314-H318-H302
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
    Chemical Name or Material Benzyltriethylammonium borohydride
    Assay 96%

    Description

    Applications
    Benzyltriethylammonium borohydride involves in the conversion of alkenes to the anti-Markovnikov alcohols in presence of TMS chloride or TiCl4. It combines with TMS chloride, selectively reduces carboxylic acids to alcohols; esters, lactones, nitro- and chloro-substituents are unaffected. It is used as reducing reagent.

    Solubility
    Souble in Chloroform.

    Notes
    Moisture sensitive. Store away from moisture and oxidizing agents under dry inert gas. Protect from humidity and water. Incompatible with oxidizing agents and moisture.

    GHS H Statement
    H261-H314-H318-H302
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit