Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzyltrimethylammonium dichloroiodate, 95%, Thermo Scientific Chemicals

Catalog Number AAB2289214
Click to view available options
Quantity:
5 g
25 g
114971-52-7
C10H16Cl2IN
348.049
MFCD00075259
PPDJNZTUDFPAHX-UHFFFAOYSA-N
benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
2724680
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl
114971-52-7
C10H16Cl2IN
348.049
MFCD00075259
PPDJNZTUDFPAHX-UHFFFAOYSA-N
benzyltrimethylammonium dichloroiodate, benzyltrimethylammonium dichloroiodide, btma-icl2, benzyl trimethylammonium dichloroiodate, benzyltrimethylazanium; dichloroiodanuide, pubchem21920, benzyltrimethylammoniumdichloroiodate
2724680
C[N+](C)(C)CC1=CC=CC=C1.Cl[I-]Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼125°C
Linear Formula C6H5CH2(CH3)3NICl2
Quantity 25 g
Beilstein 4611949
Sensitivity Hygroscopic; Light sensitive
Formula Weight 348.06
Percent Purity 95%
Chemical Name or Material Benzyltrimethylammonium dichloroiodate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit